missing translation for 'onlineSavingsMsg'
Learn More

Eriochrome™ Cyanine R, Honeywell™ Fluka™

Product Code. 15643850
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

S. No. 840

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 3564-18-9
Molecular Formula C23H15Na3O9S
MDL Number MFCD00012411
UN Number NONH for all modes of transport
Beilstein 3873874
Formula Weight 536.39g/mol
Packaging Glass bottle
Quantity 25 g
Chemical Name or Material Eriochrome™ Cyanine R
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'