Learn More
Notrax ESD Earth Bonding Plug
Click to view available options
Earth Bonding Plug; Color: Yellow
Description
- Specialized line of ESD accessories to ensure a static free workplace and protect employees and sensitive electrical equipment
- Use these accessories to connect table work surface, employees and conductive material to a common grounding point
- This creates a balance between all items and personnel to ensure static electricity will flow to ground in a controlled manner
- For use in ESD working zones to protect electrostatic sensitive components
- Connects grounding cords to ground
- 1 Meg Resistor, 3 x 10 mm male studs, allowing multiple cord connections per plug
- Available in Continental or UK version
Specifications
Specifications
Color | Yellow |
Product Type | Bounding Plug |
Material | Wire |
Connector Types | EU Plug Connection |