Learn More
Notrax ESD Non-Abrasive Mat Cleaner
Click to view available options
Mat Cleaner; For Use With: Remove Flux Residues, Oil, Finger Marks and Dirt
Description
- Mat cleaner is recommended for static safe table and floor mats
- Its non abrasive formula leaves no film and is designed to enhance the static dissipative properties of
- ESD safe surfaces
- Effectively removes flux residues, oil, finger marks and dirt
- Convenient 1000 mL bottle trigger spray
- Specialized line of ESD accessories to ensure a static free workplace and protect employees and sensitive electrical equipment
- Use these accessories to connect table work surface, employees and conductive material to a common grounding point
- This creates a balance between all items and personnel to ensure static electricity will flow to ground in a controlled manner
- For use in ESD working zones to protect electrostatic sensitive components
Specifications
Specifications
Product Type | Mat Cleaner |
Material | Wire |
For Use With (Application) | Remove Flux Residues, Oil, Finger Marks and Dirt |