missing translation for 'onlineSavingsMsg'
Learn More

Justrite ETFE Tubing Compression Fittings for HPLC Poly Manifold

ETFE Tubing Compression Fittings for HPLC Poly Manifold include ferrules for a compression fit to the waste line and are made of ETFE (ethylene tetrafluoroethylene), a fluorine based plastic

Brand:  Justrite JCN28188BU

 View more versions of this product

Product Code. 17960541

  • 1905.00 SEK / 6 units

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Sold in sets of six
  • Accommodate hard-wall tubing size of 1.6 mm OD
  • The fittings are approximately 25 mm long
  • Made of ETFE (ethylene tetrafluoroethylene), a fluorine based plastic
  • They include ferrules for a compression fit to the waste line
Specifications

Specifications

Fitting
1.6 mm
25 mm
Blue
1 in.
ETFE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.