missing translation for 'onlineSavingsMsg'
Learn More

Ethanol, Industrial, 95%, Solveco

Product Code. 11370436
Click to view available options
Quantity:
12 x 1 L
Packaging:
PET Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Completely denatured ethanol with 20g Methyl ethyl ketone and 30g Methyl isobutyl ketone. Solveco Ethanol, Industrial, 95% is part of the natural cycle. It is used extensively in the engineering industry and in washing fluids, as well as a coolant in machining.

Denatured Ethanol 95% is a clear, colorless, volatile liquid with a distinctive odor. It reacts explosively with strong oxidizing agents and can react violently with metals to produce hydrogen. It may damage gaskets, painted surfaces, protective and sealing grease coatings, materials, rubber and some plastics.

  • Fermented ethanol content of 95% by volume
  • Direct use of the bottle by attaching a spray nozzle
  • Molecular formula: CH3CH2OH
  • CAS: 64-17-5

Specifications

CAS 64-17-5
Color Colorless
Density 0.8109–0.8119kg/L
Quantity 12 x 1 L
Packaging PET Bottle
Molecular Formula C2H6O
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILES CCO
IUPAC Name ethanol
Molecular Weight (g/mol) 46.069
PubChem CID 702
ChEBI CHEBI:16236
Formula Weight 46.07g/mol
Percent Purity 92.4% ±2% w/w
Show More Show Less
compliance-icons
Product Identifier
  • Industri Etanol 95% (denaturerad med MIBK och MEK)
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contains : Metyletylketon, Etanol, Metylisobutylketon
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.