missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-(4-hydroxyphenyl)-4-methylthiazole-5-carboxylate, 97%, Thermo Scientific™

Product Code. 15439665
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 161797-99-5
Molecular Formula C13H13NO3S
Molecular Weight (g/mol) 263.31
MDL Number MFCD03700424
InChI Key VBAMDWNNTNVLAV-UHFFFAOYSA-N
PubChem CID 7172170
IUPAC Name ethyl 4-methyl-2-(4-oxocyclohexa-2,5-dien-1-ylidene)-3H-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)C1=C(C)NC(S1)=C1C=CC(=O)C=C1

Specifications

Quantity 5 g
Synonym ethyl 2-4-hydroxyphenyl-4-methylthiazole-5-carboxylate, ethyl2-4-hydroxyphenyl-4-methylthiazole-5-carboxylate, ethyl 2-4-hydroxyphenyl-4-methyl thiazole-5-carboxylate, ethyl 2-4-hydroxyphenyl-4-methyl-1,3-thiazole-5-carboxylate, 2-4-hydroxyphenyl-4-methyl-thiazole-5-carboxylic acid ethyl ester, 5-thiazolecarboxylic acid, 2-4-hydroxyphenyl-4-methyl-, ethyl ester, 2-4-hydroxyphenyl-4-methylthiazole-5-carboxylate, ethyl 2-4-hydroxyphenyl-4-methyl-5-thiazolecarboxylate, 2-4-hydroxy-phenyl-4-methyl-thiazole-5-carboxylic acid ethyl ester, 2-4-hydroxyphenyl-4-methyl-thiazole-5-carboxylicacid ethyl ester
Formula Weight 263.31
Percent Purity 97%
Chemical Name or Material Ethyl 2-(4-hydroxyphenyl)-4-methylthiazole-5-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'