Learn More
Ethyl 3,5-dimethyl-1H-4-pyrazolecarboxylate, 97%, Thermo Scientific™
602.61 SEK - 3443.74 SEK
Chemical Identifiers
CAS | 35691-93-1 |
---|---|
Molecular Formula | C8H12N2O2 |
Molecular Weight (g/mol) | 168.196 |
MDL Number | MFCD00159638 |
InChI Key | BCKARVLFIJPHQU-UHFFFAOYSA-N |
Synonym | ethyl 3,5-dimethyl-1h-4-pyrazolecarboxylate, ethyl 3,5-dimethylpyrazole-4-carboxylate, 3,5-dimethylpyrazole-4-carboxylic acid ethyl ester, 3,5-dimethyl-1h-pyrazole-4-carboxylic acid ethyl ester, 4-pyrazolecarboxylic acid, 3,5-dimethyl-, ethyl ester, 3.5. dimethylpyrazol 4-carbonsaeureaethyl ester german, 3.5. dimethylpyrazol 4-carbonsaeureaethyl ester, 3,5-dimethyl-4-ethoxycarbonyl-1h-pyrazole, 1h-pyrazole-4-carboxylic acid, 3,5-dimethyl-, ethyl ester, deeShow More |
PubChem CID | 215436 |
IUPAC Name | ethyl 3,5-dimethyl-1H-pyrazole-4-carboxylate |
SMILES | CCOC(=O)C1=C(NN=C1C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
10595753
|
Thermo Scientific Maybridge
CC01223DA |
1 g |
602.61 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
10278032
|
Thermo Scientific Maybridge
CC01223EA |
10 g | N/A | N/A | N/A | |||||
Description
Chemical Identifiers
35691-93-1 | |
168.196 | |
BCKARVLFIJPHQU-UHFFFAOYSA-N | |
215436 | |
CCOC(=O)C1=C(NN=C1C)C |
C8H12N2O2 | |
MFCD00159638 | |
ethyl 3,5-dimethyl-1h-4-pyrazolecarboxylate, ethyl 3,5-dimethylpyrazole-4-carboxylate, 3,5-dimethylpyrazole-4-carboxylic acid ethyl ester, 3,5-dimethyl-1h-pyrazole-4-carboxylic acid ethyl ester, 4-pyrazolecarboxylic acid, 3,5-dimethyl-, ethyl ester, 3.5. dimethylpyrazol 4-carbonsaeureaethyl ester german, 3.5. dimethylpyrazol 4-carbonsaeureaethyl ester, 3,5-dimethyl-4-ethoxycarbonyl-1h-pyrazole, 1h-pyrazole-4-carboxylic acid, 3,5-dimethyl-, ethyl ester, deeShow More | |
ethyl 3,5-dimethyl-1H-pyrazole-4-carboxylate |
Specifications
35691-93-1 | |
100.0 | |
C8H12N2O2 | |
ethyl 3,5-dimethyl-1h-4-pyrazolecarboxylate, ethyl 3,5-dimethylpyrazole-4-carboxylate, 3,5-dimethylpyrazole-4-carboxylic acid ethyl ester, 3,5-dimethyl-1h-pyrazole-4-carboxylic acid ethyl ester, 4-pyrazolecarboxylic acid, 3,5-dimethyl-, ethyl ester, 3.5. dimethylpyrazol 4-carbonsaeureaethyl ester german, 3.5. dimethylpyrazol 4-carbonsaeureaethyl ester, 3,5-dimethyl-4-ethoxycarbonyl-1h-pyrazole, 1h-pyrazole-4-carboxylic acid, 3,5-dimethyl-, ethyl ester, dee | |
CCOC(=O)C1=C(NN=C1C)C | |
168.196 | |
168.19 | |
Ethyl 3, 5-dimethyl-1H-4-pyrazolecarboxylate |
96.5 | |
97% | |
MFCD00159638 | |
BCKARVLFIJPHQU-UHFFFAOYSA-N | |
ethyl 3,5-dimethyl-1H-pyrazole-4-carboxylate | |
215436 | |
97% |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.