Learn More
Ethyl 4-nitrophenylacetate, 98%, Thermo Scientific Chemicals
694.00 SEK - 7180.00 SEK
Chemical Identifiers
CAS | 5445-26-1 |
---|---|
Molecular Formula | C10H11NO4 |
Molecular Weight (g/mol) | 209.20 |
MDL Number | MFCD00017046 |
InChI Key | DWDRNKYLWMKWTH-UHFFFAOYSA-N |
Synonym | ethyl 4-nitrophenylacetate, ethyl 2-4-nitrophenyl acetate, ethyl p-nitrophenylacetate, ethyl 4-nitrobenzeneacetate, ethyl 4-nitrophenyl acetate, 4-nitrophenylacetic acid ethyl ester, benzeneacetic acid, 4-nitro-, ethyl ester, ethyl-4-nitrophenylacetate, acetic acid, p-nitrophenyl-, ethyl ester, 4-nitro-phenyl-acetic acid ethyl ester |
PubChem CID | 79517 |
IUPAC Name | ethyl 2-(4-nitrophenyl)acetate |
SMILES | CCOC(=O)CC1=CC=C(C=C1)[N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11418983
|
Thermo Scientific Alfa Aesar
A11101.06 |
5 g |
694.00 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11428983
|
Thermo Scientific Alfa Aesar
A11101.14 |
25 g |
2135.00 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11438983
|
Thermo Scientific Alfa Aesar
A11101.22 |
100 g |
7180.00 SEK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5445-26-1 | |
209.20 | |
DWDRNKYLWMKWTH-UHFFFAOYSA-N | |
79517 | |
CCOC(=O)CC1=CC=C(C=C1)[N+]([O-])=O |
C10H11NO4 | |
MFCD00017046 | |
ethyl 4-nitrophenylacetate, ethyl 2-4-nitrophenyl acetate, ethyl p-nitrophenylacetate, ethyl 4-nitrobenzeneacetate, ethyl 4-nitrophenyl acetate, 4-nitrophenylacetic acid ethyl ester, benzeneacetic acid, 4-nitro-, ethyl ester, ethyl-4-nitrophenylacetate, acetic acid, p-nitrophenyl-, ethyl ester, 4-nitro-phenyl-acetic acid ethyl ester | |
ethyl 2-(4-nitrophenyl)acetate |
Specifications
5445-26-1 | |
C10H11NO4 | |
2053241 | |
DWDRNKYLWMKWTH-UHFFFAOYSA-N | |
ethyl 2-(4-nitrophenyl)acetate | |
79517 | |
98% |
62°C to 66°C | |
MFCD00017046 | |
ethyl 4-nitrophenylacetate, ethyl 2-4-nitrophenyl acetate, ethyl p-nitrophenylacetate, ethyl 4-nitrobenzeneacetate, ethyl 4-nitrophenyl acetate, 4-nitrophenylacetic acid ethyl ester, benzeneacetic acid, 4-nitro-, ethyl ester, ethyl-4-nitrophenylacetate, acetic acid, p-nitrophenyl-, ethyl ester, 4-nitro-phenyl-acetic acid ethyl ester | |
CCOC(=O)CC1=CC=C(C=C1)[N+]([O-])=O | |
209.20 | |
209.2 | |
Ethyl 4-nitrophenylacetate |
Safety and Handling
missing translation for 'einecsNumber' : 226-646-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.