missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-methylisoxazole-4-carboxylate, 96%, Thermo Scientific™

310.95 SEK - 1029.72 SEK

Chemical Identifiers

CAS 51135-73-0
Molecular Formula C7H9NO3
Molecular Weight (g/mol) 155.153
MDL Number MFCD01631119
InChI Key KOMSQTMQKWSQDW-UHFFFAOYSA-N
Synonym ethyl 5-methylisoxazole-4-carboxylate, 5-methyl-4-isoxazolecarboxylate, ethyl 5-methyl-4-isoxazolecarboxylate, 5-methylisoxazole-4-carboxylic acid ethyl ester, acmc-20aky5, ethyl5-methylisoxazole-4-carboxylate, ethyl-5-methylisoxazole-4-carboxylate, ethyl 5-methylisoxazol-4-yl carboxylate, ethyl 5-m+c3117ethyl-4-isoxazolecarboxylate
PubChem CID 7009317
IUPAC Name ethyl 5-methyl-1,2-oxazole-4-carboxylate
SMILES CCOC(=O)C1=C(ON=C1)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11349435
SDS View Documents
Thermo Scientific Alfa Aesar
L18647.04
2 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11359435
SDS View Documents
Thermo Scientific Alfa Aesar
L18647.09
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

51135-73-0
155.153
KOMSQTMQKWSQDW-UHFFFAOYSA-N
7009317
CCOC(=O)C1=C(ON=C1)C
C7H9NO3
MFCD01631119
ethyl 5-methylisoxazole-4-carboxylate, 5-methyl-4-isoxazolecarboxylate, ethyl 5-methyl-4-isoxazolecarboxylate, 5-methylisoxazole-4-carboxylic acid ethyl ester, acmc-20aky5, ethyl5-methylisoxazole-4-carboxylate, ethyl-5-methylisoxazole-4-carboxylate, ethyl 5-methylisoxazol-4-yl carboxylate, ethyl 5-m+c3117ethyl-4-isoxazolecarboxylate
ethyl 5-methyl-1,2-oxazole-4-carboxylate

Specifications

51135-73-0
C7H9NO3
MFCD01631119
ethyl 5-methylisoxazole-4-carboxylate, 5-methyl-4-isoxazolecarboxylate, ethyl 5-methyl-4-isoxazolecarboxylate, 5-methylisoxazole-4-carboxylic acid ethyl ester, acmc-20aky5, ethyl5-methylisoxazole-4-carboxylate, ethyl-5-methylisoxazole-4-carboxylate, ethyl 5-methylisoxazol-4-yl carboxylate, ethyl 5-m+c3117ethyl-4-isoxazolecarboxylate
CCOC(=O)C1=C(ON=C1)C
155.153
155.15
Ethyl 5-methylisoxazole-4-carboxylate
95°C to 97°C (15 mmHg)
1.4597
122097
KOMSQTMQKWSQDW-UHFFFAOYSA-N
ethyl 5-methyl-1,2-oxazole-4-carboxylate
7009317
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

missing translation for 'einecsNumber' : 000-000-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl 5-methylisoxazole-4-carboxylate, 96%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.