missing translation for 'onlineSavingsMsg'
Learn More

Ethyl coumarin-3-carboxylate, 98%, Thermo Scientific Chemicals

331.00 SEK - 1015.00 SEK

Chemical Identifiers

CAS 1846-76-0
Molecular Formula C12H10O4
Molecular Weight (g/mol) 218.208
MDL Number MFCD00016964
InChI Key XKHPEMKBJGUYCM-UHFFFAOYSA-N
Synonym ethyl coumarin-3-carboxylate, ethyl 2-oxo-2h-chromene-3-carboxylate, 3-ethoxycarbonylcoumarin, ethyl 3-coumarincarboxylate, 3-carbethoxycoumarin, ethylcoumarin-3-carboxylate, coumarin-3-carboxylic acid ethyl ester, 1,2-benzopyran-2-one-3-carboxylic acid, ethyl ester, 2h-1-benzopyran-3-carboxylic acid, 2-oxo-, ethyl ester, 2h-1-benzopyran-3-carboxylicacid, 2-oxo-, ethyl ester
PubChem CID 15800
IUPAC Name ethyl 2-oxochromene-3-carboxylate
SMILES CCOC(=O)C1=CC2=CC=CC=C2OC1=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11404254
SDS View Documents
Thermo Scientific Alfa Aesar
A12553.03
1 g
331.00 SEK
1g
Please to purchase this item. Need a web account? Register with us today!
11414254
SDS View Documents
Thermo Scientific Alfa Aesar
A12553.06
5 g
1015.00 SEK
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'rtecsNumber' : DJ2501000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl coumarin-3-carboxylate, 98%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.