Learn More
Ethyl hydrogen suberate, 98%, Thermo Scientific Chemicals
Product Code. 11366866
Click to view available options
Quantity:
5 g
25 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 14113-01-0
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00014385
InChI Key KCFVQEAPUOVXTH-UHFFFAOYSA-N
PubChem CID 84204
IUPAC Name 8-ethoxy-8-oxooctanoic acid
SMILES CCOC(=O)CCCCCCC(=O)O
Specifications
Melting Point 25°C to 27°C
Quantity 25 g
Beilstein 1775737
Synonym ethyl hydrogen suberate, octanedioic acid, ethyl, korksaureathylester, acmc-1b2e5, 8-ethoxy-8-oxo-octanoic acid, octanedioic acid 1-ethyl ester, octanedioic acid,1-ethyl ester, octanedioic acid, 1-ethyl ester, 8-ethoxy-8-oxidanylidene-octanoic acid, ethylhydrogensuberate, monoethylsuberate; subericacid monoethylester
Formula Weight 202.25
Percent Purity 98%
Chemical Name or Material Ethyl hydrogen suberate