missing translation for 'onlineSavingsMsg'
Learn More

Ethyl piperidine-1-acetate, 98%, Thermo Scientific™

Product Code. 11460254
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23853-10-3
Molecular Formula C9H17NO2
Molecular Weight (g/mol) 171.24
MDL Number MFCD00134496
InChI Key LXZHLNWWDDNIOJ-UHFFFAOYSA-N
PubChem CID 643466
IUPAC Name ethyl 2-piperidin-1-ylacetate
SMILES CCOC(=O)CN1CCCCC1

Specifications

Density 0.984
Boiling Point 109°C to 111°C (25 mmHg)
Flash Point 91°C (195°F)
Refractive Index 1.454
Quantity 5 g
Beilstein 123142
Synonym ethyl 1-piperidineacetate, ethyl piperidin-1-ylacetate, ethyl 2-piperidin-1-yl acetate, ethyl piperidine-1-acetate, 1-piperidineacetic acid, ethyl ester, ethyl 2-piperidylacetate, ethyl1-piperidineacetate, acmc-20al1z, ethyl 1-piperidine acetate, ethyl 2-piperidinylacetate
Formula Weight 171.24
Percent Purity 98%
Chemical Name or Material Ethyl piperidine-1-acetate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'