missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ethyle1-methylcyclopropane-1-carboxylate, 97%, Thermo Scientific™
344.00 SEK - 3590.00 SEK
Chemical Identifiers
CAS | 71441-76-4 |
---|---|
Molecular Formula | C7H12O2 |
Molecular Weight (g/mol) | 128.171 |
MDL Number | MFCD00001284 |
InChI Key | IZPYNZLFBUQGCZ-UHFFFAOYSA-N |
PubChem CID | 544234 |
IUPAC Name | ethyl 1-methylcyclopropane-1-carboxylate |
SMILES | CCOC(=O)C1(CC1)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10293462
|
Thermo Scientific Maybridge
SB01040DA |
1 g |
344.00 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10794383
|
Thermo Scientific Maybridge
SB01040EA |
10 g |
1850.00 SEK
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10440764
|
Thermo Scientific Maybridge
SB01040EB |
25 g |
3590.00 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
71441-76-4 | |
128.171 | |
IZPYNZLFBUQGCZ-UHFFFAOYSA-N | |
ethyl 1-methylcyclopropane-1-carboxylate |
C7H12O2 | |
MFCD00001284 | |
544234 | |
CCOC(=O)C1(CC1)C |
Specifications
71441-76-4 | |
0.9180g/mL | |
Amber Glass Bottle | |
CH3C3H4C(O)OCH2CH3 | |
ethyl 1-methylcyclopropanecarboxylate, 1-methylcyclopropanecarboxylic acid ethyl ester, ethyl1-methylcyclopropanecarboxylate, cyclopropanecarboxylic acid, 1-methyl-, ethyl ester, cyclopropanecarboxylicacid, 1-methyl-, ethyl ester, ethyl 1-methylcyclopropane-carboxylate, 1-methylcyclopropanoic acid ethyl ester, 1-ethoxycarbonyl-1-methylcyclopropane, ethyl 1-methylcyclopropan-1-carboxylate, ethyl 1-methyl cyclopropane-1-carboxylate | |
CCOC(=O)C1(CC1)C | |
128.171 | |
128.17 | |
Ethyl 1-methylcyclopropane-1-carboxylate |
97.0 | |
97% | |
C7H12O2 | |
MFCD00001284 | |
IZPYNZLFBUQGCZ-UHFFFAOYSA-N | |
ethyl 1-methylcyclopropane-1-carboxylate | |
544234 | |
97% |
Safety and Handling
GHS H Statement:
Flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.