Learn More
Ethyl methanesulfonate, 99%, Thermo Scientific Chemicals
1230.71 SEK - 27312.32 SEK
Chemical Identifiers
CAS | 62-50-0 |
---|---|
Molecular Formula | C3H8O3S |
Molecular Weight (g/mol) | 124.15 |
MDL Number | MFCD00007559 |
InChI Key | PLUBXMRUUVWRLT-UHFFFAOYSA-N |
Synonym | ethyl mesylate, ethyl methanesulphonate, half-myleran, methanesulfonic acid, ethyl ester, methanesulfonic acid ethyl ester, methylsulfonic acid, ethyl ester, rcra waste number u119, ethyl methansulphonate, ethyl ester of methylsulfonic acid, ethyl ester of methanesulfonic acid |
PubChem CID | 6113 |
ChEBI | CHEBI:23994 |
IUPAC Name | ethyl methanesulfonate |
SMILES | CCOS(=O)(=O)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10521461
|
Thermo Scientific Acros
205260100 |
10 g | Glass bottle |
1230.71 SEK
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10398050
|
Thermo Scientific Acros
205260250 |
25 g | Glass bottle |
2201.08 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10145643
|
Thermo Scientific Acros
205261000 |
100 g | Glass bottle |
6532.24 SEK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11934561
|
Thermo Scientific Acros
205265000 |
500 g | Glass bottle |
27312.32 SEK
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
62-50-0 | |
124.15 | |
PLUBXMRUUVWRLT-UHFFFAOYSA-N | |
6113 | |
ethyl methanesulfonate |
C3H8O3S | |
MFCD00007559 | |
ethyl mesylate, ethyl methanesulphonate, half-myleran, methanesulfonic acid, ethyl ester, methanesulfonic acid ethyl ester, methylsulfonic acid, ethyl ester, rcra waste number u119, ethyl methansulphonate, ethyl ester of methylsulfonic acid, ethyl ester of methanesulfonic acid | |
CHEBI:23994 | |
CCOS(=O)(=O)C |
Specifications
62-50-0 | |
213.0°C to 214.0°C | |
99% | |
CH3SO3C2H5 | |
MFCD00007559 | |
15,3882 | |
ethyl mesylate, ethyl methanesulphonate, half-myleran, methanesulfonic acid, ethyl ester, methanesulfonic acid ethyl ester, methylsulfonic acid, ethyl ester, rcra waste number u119, ethyl methansulphonate, ethyl ester of methylsulfonic acid, ethyl ester of methanesulfonic acid | |
PLUBXMRUUVWRLT-UHFFFAOYSA-N | |
ethyl methanesulfonate | |
6113 | |
124.15 | |
Ethyl methanesulfonate |
1.1600g/mL | |
100°C | |
C3H8O3S | |
1.4170 to 1.4190 | |
04,5 | |
1.16 | |
Solubility in water: 50-100g/L (27°C) | |
CCOS(=O)(=O)C | |
124.15 | |
CHEBI:23994 | |
99% |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
May cause genetic defects.
May cause cancer.
Causes skin irritation.
Harmful if swallowed.
GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-536-7
- Ethyl methanesulfonate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 1A
- Germ cell mutagenicity Category 1A
Hazard Statement
- H340-May cause genetic defects.
- H350-May cause cancer.
- H302-Harmful if swallowed.
Precautionary Statement
- P201-Obtain special instructions before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only