missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic acid, 99%, Thermo Scientific Chemicals

739.00 SEK - 3395.00 SEK

Chemical Identifiers

CAS 60-00-4
Molecular Formula C10H16N2O8
Molecular Weight (g/mol) 292.24
MDL Number MFCD00003541
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
Synonym edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
PubChem CID 6049
ChEBI CHEBI:42191
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11408920
SDS View Documents
Thermo Scientific Alfa Aesar
A10713.36
500 g
739.00 SEK
500g
Please to purchase this item. Need a web account? Register with us today!
11479562
SDS View Documents
Thermo Scientific Alfa Aesar
A10713.0B
1000 g
895.00 SEK
1000g
Please to purchase this item. Need a web account? Register with us today!
11499562
SDS View Documents
Thermo Scientific Alfa Aesar
A10713.0I
5000 g
3395.00 SEK
5000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ethylenediaminetetraacetic acid is used as a complexing agent for many cations. It prevents metal ion impurities and used to remove excess iron from the body. It acts as a sequestering agent in cosmetic products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylenediaminetetraacetic acid is used as a complexing agent for many cations. It prevents metal ion impurities and used to remove excess iron from the body. It acts as a sequestering agent in cosmetic products.

Solubility
Soluble in sodium hydroxide, sodium carbonate, ammonia solution, acid and common organic solvents. Slightly soluble in water.

Notes
Stable. Incompatible with copper, copper alloys, nickel, aluminum, strong oxidizing agents and strong bases.
Specifications

Chemical Identifiers

60-00-4
292.24
KCXVZYZYPLLWCC-UHFFFAOYSA-N
6049
2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid
C10H16N2O8
MFCD00003541
edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
CHEBI:42191
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

60-00-4
C10H16N2O8
MFCD00003541
14,3517
Soluble in sodium hydroxide,sodium carbonate,ammonia solution,acid and common organic solvents. Slightly soluble in water.
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
292.24
CHEBI:42191
99%
∼245°C (decomposition)
(HO2CCH2)2NCH2CH2N(CH2CO2H)2
1716295
edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
KCXVZYZYPLLWCC-UHFFFAOYSA-N
2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid
6049
292.24
Ethylenediaminetetraacetic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

P260-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P314-P501c

H319-H332-H373

missing translation for 'einecsNumber' : 200-449-4

missing translation for 'rtecsNumber' : AH4025000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Ethylenediaminetetraacetic acid

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Ethylenediamine tetraacetic acid

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethylenediaminetetraacetic acid, 99%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.