missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediaminetetraacetic Acid Trisodium Salt Monohydrate, 99.32%, MP Biomedicals™
Click to view available options
Quantity:
100 g
Specifications
Specifications
CAS | 10378-22-0 |
Color | White |
Molecular Formula | C10H18N2Na3O9 |
MDL Number | MFCD00149675 |
Quantity | 100 g |
Synonym | edta, trisodium salt trihydrate |
InChI Key | DFCJTQJARZUURB-UHFFFAOYSA-N |
SMILES | C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.O.[Na].[Na].[Na] |
IUPAC Name | 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;sodium;hydrate |
Molecular Weight (g/mol) | 379.228 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'