Learn More
Invitrogen™ EVOS™ Light Cube, QDOT WB 525-800 2.0
EVOS LED light cubes provide superior illumination uniformity across the field of view, resulting in exceptional tile-stitch image quality. At the same time, optimized spectral fidelity across channels significantly improves data quality by minimizing undesired bleed-through.
Brand:  Invitrogen™ AMEP4966
46341.00 SEK valid until 2025-03-31
Use promo code "24119" to get your promotional price.
Product Code. 17344493
  • 48780.00 SEK / Each
Estimated Shipment: 07-03-2025
to see stock.
Explore more special offers
Description
  • Second-generation EVOS LED light cubes provide superior illumination uniformity across the field of view, resulting in exceptional tile-stitch image quality
  • At the same time, optimized spectral fidelity across channels significantly improves data quality by minimizing undesired bleed-through
  • Like the original design, they combine condensing optics and hard-coated filters into one light engine that delivers digitally controlled fluorophore excitation
  • These LED light cubes are backwards compatible with older EVOS and Countess systems. and can be used with first-generation light cubes
  • Like the original light cubes, these LED cubes are user-changeable and automatically recognized by the instrument
  • The EVOS Light Cube, QDOT WB 525-800 2.0, is ideal for use with Qdot fluorophores, and other fluorophores with excitation and emission maxima near 445 and 514 nm
  • The LED bulbs on the EVOS systems are rated for >50,000 hours, compared to 300 hours for a typical mercury bulb and 1,500 hours for a metal halide bulb
EVOS LED light cubes offer these important advantages:
  • Uniform illumination across the entire field of view for superb image quality
  • Optimized spectral compatibility to maximize true signal detection, while minimizing bleed-through for the best possible data quality
  • A short light path that affords superior detection of fluorescent signals
  • Stable illumination intensity over 50,000 hours that helps experimental reproducibility
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 each