Learn More
EVOS™ EVOS™ Condenser Slider, Pinhole
Adds contrast when imaging
Brand: EVOS™ AMEPAS15
Additional Details : Weight : 2.00000kg
Description
- Used to improve illumination under certain conditions
- Due to variances in sample size, color and thickness, actual slider use may differ from suggestions
- Intended for brightfield applications at all magnifications to enhance contrast
Pinhole condenser slider is included with the EVOS™ FL and FL Color imaging systems.
Specifications
Condenser Slider | |
EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System | |
1 Ea. |
EVOS | |
Diffuser Slider, Pinhole Slider, Meniscus Sliders |
For Research Use Only. Not for use in diagnostic procedures.