Learn More
Thermo Scientific™ EVOS™ Power Cord, Type I (Australia)
This power cord is for use with EVOS XL, FL , FL Color, and FL Auto systems in countries with Type I power plugs.
Brand: Thermo Scientific™ AMEP4647
Additional Details : Weight : 0.50000kg
Description
This power cord is for use with EVOS XL, FL , FL Color, and FL Auto systems in countries with Type I power plugs. The Type I electrical plug is mainly used in Australia and New Zealand, but is also used in China and parts of Asia, South America, and Africa.
Specifications
Power Cord | |
Type I (Electrical Plug), Type I (Power Plug) | |
1 Ea. |
EVOS | |
EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System |