Learn More
EVOS™ Stage Plate Adapter
EVOS™ Stage Plate Adapter
Brand: Invitrogen AMEP4692
Additional Details : Weight : 2.00000kg
Description
The EVOS™ Stage Plate Adapter is designed to fit into the EVOS™ Staging Plate with 160 X 110 mm opening. This adapter accommodates vessel holders AMEPVH001 through AMEPVH018 for precision in sample alignment. With a snug and secure fit, yet readily switched, the plate/adapter combination make imaging samples in a range of vessel holders convenient and easy.For additional choices, see the EVOS™ Guide to Vessel Holders and Stage Plates.
Explore the entire EVOS™ line of imaging systems and accessories.
Specifications
Stage Plate Adapter | |
EVOS™ Stage Plate Adapter, 128.2 L x 86.2mm W Holding Size, With a Snug and Secure Fit, Use with EVOS™ FL Color Imaging System, EVOS™ FL Imaging System, EVOS™ XL Imaging System, Cellular Imaging | |
EVOS™ FL Color Imaging System, EVOS™ FL Imaging System, EVOS™ XL Imaging System | |
1 Holder |
For Research Use Only. Not for use in diagnostic procedures.