Learn More
Invitrogen™ EVOS™ Universal Stage Plate
This EVOS Universal Stage Plate is designed to make a quick check of samples fast and easy.
Brand:  Invitrogen™ AMEPVH065
Additional Details : Weight : 0.68000kg
Product Code. 16366602
  • 3010.00 SEK / Each
Estimated Shipment: 07-08-2024
to see stock.
Add to basket
Description
Description
With a 1.5-inch diameter opening, it enables you to quickly observe cell staining, GFP expression, cell morphology, or confluence by simply moving the sample vessel across the stage. No additional vessel holders or accessories are needed. When more precise and secure alignment of your sample is desired, use a vessel holder from our wide range of holders for microscope slides, cell culture flasks, Petri dishes, microwell plates, and more that are accommodated on the EVOS microscope in conjunction with a stage plate such as Cat. Nos. AMEPVH089 or AMEPVH054.
The EVOS Universal Stage Plate offers these advantages:
• Exceptional flexibility
• Quick imaging by simply moving the vessel across the stage
• Compatible vessel holder adapters for a range of vessels holders
Specifications
Specifications
Universal Stage Plate
EVOS
Cell Culture Flask, Evos Microscope, Microscope Slide, Microwell Plate, Petri Dish
Room Temperature
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only. Not for use in diagnostic procedures.