Learn More
EVOS™ EVOS™ Vessel Holders Pack
Provides snug and secure fit of microscope slide, cell culture flask or dish, or micro-well plate to the stage of the EVOS imaging systems
Brand: EVOS™ AMEP4619
Additional Details : Weight : 0.01000kg
Description
- Provides increased precision in sample alignment
- Easily switchable vessel holders for convenient and easy imaging of a range of samples
- Includes 10 most popular EVOS vessel holders
- For use with: EVOS XL Core Imaging System, EVOS FL Auto Imaging System, EVOS XL Imaging System, EVOS FL Imaging System, EVOS FL Color Imaging System
Specifications
Vessel Holder | |
EVOS | |
EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System | |
T-25 flask |
For Research Use Only. Not for use in diagnostic procedures.