Learn More
Memmert™ Extended Overtemperature Protection for SN260
Extended overtemperature protection with integrated Pt100 sensor for independent temperature monitoring.
Brand:  Memmert™ SN260A6
Product Code. 17598525
  • 2120.00 SEK / Each
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Description
  • Accessory
  • With integrated Pt100 sensor
  • For models with SingleDISPLAY
Specifications
Specifications
SN260 Sterilizer
Integrated Pt100 Sensor