Learn More
Fisherbrand™ External Probe
For Use With: Isotemp (±1.8°C at 70°C) and Isotemp Advanced (±1.2°C at 70°C) Hotplates and Hotplate Stirrers
Brand: Fisherbrand™ CIC0001465
605.74 SEK valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Description
- Optimize performance and flexibility
Specifications
Isotemp (±1.8°C at 70°C) and Isotemp Advanced (±1.2°C at 70°C) Hotplates and Hotplate Stirrers | |
0°C to 400°C | |
Probe |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
23Hours
1Minutes
10Seconds
18 -
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now