Learn More
Grant Instruments™ External Probe
External probes for monitoring and controlling temperature of remote loads.
Brand:  Grant Instruments™ SEP
 View more versions of this product
Product Code. 15449224
  • 2775.00 SEK / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
Grant Instruments temperature probe (Pt1000) external for use with LT ecocool 150
  • 3 m cable
Specifications
Specifications
Stainless Steel
3 m