Learn More
Lenz Laborglasintrumente Extractor Thimbles for Solids
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 01506029
This item is not returnable.
View return policy
Description
- Expansion adapters
- With ground joints
- Acc. to DIN 12257
- Conical ground sockets and cones
- Made of DURAN™ tubing
Specifications
NS 29/32 | |
Borosilicate Glass (3.3) | |
NS 60/46 Socket |