Learn More
Lenz Laborglasintrumente Extractors for Solids
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
2300.20 SEK - 3248.90 SEK
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10060521
|
Lenz Laborglasintrumente
05431049 |
250 mL |
3248.90 SEK
Each |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||||
10356121
|
Lenz Laborglasintrumente
05431031 |
70 mL |
2300.20 SEK
Each |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||||
10488402
|
Lenz Laborglasintrumente
05431037 |
100 mL |
2337.30 SEK
Each |
Estimated Shipment: 11-10-2024
Log in to see stock. |
|||||
Description
- Extractor head with glass thimbles for solids
- Made of DURAN™ tubing
- Dimroth condenser (details on page 71)
- With thread GL 14
- Round bottom flask
Specifications
Borosilicate Glass 3.3 | |
NS 45/40 |