missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F(ab')2-Donkey anti-Rat IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Invitrogen™
F(ab')2-Donkey Polyclonal Secondary Antibody
Brand: Invitrogen A24545
This item is not returnable.
View return policy
Description
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Rat IgG (H+L) Highly Cross-Adsorbed | |
Polyclonal | |
Unconjugated | |
Donkey | |
Affinity Chromatography | |
RUO | |
Rat | |
Liquid |
ELISA | |
> 4.5 mg/mL | |
PBS with 0.05% sodium azide; pH 7.2 | |
Gamma Immunoglobins Heavy and Light chains | |
1 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |