Learn More
Memmert™ Factory Calibration (3 Temperatures) for UF160plus
Calibration at Memmert factory for three temperature value.
Brand: Memmert™ UF160plusD00128
Description
- For three temperature values: 100°C, 160°C, and 220°C
Specifications
UF160plus Oven | |
Measuring point chamber center | |
Certificate for 3 temperatures | |
Standard Works Calibration Certificate |