missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FADS1 Rabbit anti-Human, Mouse, Rat, Clone: 1M8L2, Novus Biologicals™
Rabbit Monoclonal Antibody
1905.00 SEK - 4805.00 SEK
Specifications
Antigen | FADS1 |
---|---|
Clone | 1M8L2 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18322405
|
Bio-Techne
NBP3-15253-20UL |
20 μg |
1905.00 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18382804
|
Bio-Techne
NBP3-15253-100UL |
100 μg |
4805.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FADS1 Monoclonal antibody specifically detects FADS1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
FADS1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Diabetes Research, Lipid and Metabolism | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
3992 | |
IgG | |
Affinity purified |
1M8L2 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
D5DFLJ90273, Delta(5) desaturase, Delta(5) fatty acid desaturase, Delta-5 desaturase, delta-5 fatty acid desaturase, EC 1.14.19, EC 1.14.19.-, FADS6, FADSD5LLCDL1, fatty acid desaturase 1, FLJ38956, linoleoyl-CoA desaturase (delta-6-desaturase)-like 1, TU12 | |
A synthetic peptide corresponding to a sequence within amino acids 50-150 of human FADS1 (O60427). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title