missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FADS3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15205-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the fatty acid desaturase (FADS) gene family. Desaturase enzymes regulate unsaturation of fatty acids through the introduction of double bonds between defined carbons of the fatty acyl chain. FADS family members are considered fusion products composed of an N-terminal cytochrome b5-like domain and a C-terminal multiple membrane-spanning desaturase portion, both of which are characterized by conserved histidine motifs. This gene is clustered with family members FADS1 and FADS2 at 11q12-q13.1; this cluster is thought to have arisen evolutionarily from gene duplication based on its similar exon/intron organization.Specifications
FADS3 | |
Polyclonal | |
Unconjugated | |
FADS3 | |
CYB5RP, Cytochrome b5 related protein, FADS3, fatty acid desaturase 3, LLCDL3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
286922, 3995, 60527 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8K1P9, Q9JJE7, Q9Y5Q0 | |
Fads3 | |
FADS3 Fusion Protein Ag7355 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |