Learn More
Thermo Scientific™ FastRuler Low Range DNA Ladder, Ready-to-Use, 50-1500 bp PROMO
Perform fast sizing and approximate quantification of dsDNA in high throughput gels and conventional agarose gels with Thermo Scientific™ FastRuler™ DNA Ladders.
Brand:  Thermo Scientific™ SM1103-RMD123
Product Code. 15699484
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
Highlights
  • Fast separation (8 to 14 min)
  • Short separation distance (10 to 20 mm)
  • Sharp bands, easy-to-remember fragment sizes and quantities
  • Ready-to-use -premixed with loading dye
  • Supplied with loading dye solution for sample DNA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
6X Loading Dye
DNA Molecular Weight Marker/Ladder
Liquid
1 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ FastRuler Low Range DNA Ladder, Ready-to-Use, 50-1500 bp PROMO >
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
This product or its use is covered by at least one claim of U.S. Patent Nos. 5,834,201, 6,680,378, and/or 7,132,520 owned by Invitrogen Corporation. The purchase of this product conveys to the buyer the non-transferable right to use the purchased amount of the product in internal research conducted by the buyer. The buyer cannot use this product or materials made by the employment of this product for Commercial Purposes. Commercial Purposes means any activity for consideration and may include, but is not limited to: (1) use of the product in manufacturing; (2) use of the product to provide a service, information, or data; (3) use of the product for therapeutic, diagnostic or prophylactic purposes; or (4) resale of the product, whether or not the product is resold for use in research. Further information on purchasing licenses under the above patents may be obtained by contacting the Business Development Department, Invitrogen Corporation, 5791 Van Allen Way, Carlsbad, CA 92008.