missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferroin indicator solution, Honeywell Fluka™
Redox Indicator, Reag. Ph. Eur., E0 in sulfuric acid 1 mol/l = +1,06 volt
881.14 SEK - 9445.07 SEK
Chemical Identifiers
CAS | 14634-91-4 |
---|---|
Molecular Formula | C36H24FeN6O4S |
Molecular Weight (g/mol) | 692.53 |
MDL Number | MFCD00036428 |
InChI Key | CIWXFRVOSDNDJZ-UHFFFAOYSA-L |
Synonym | ferroin, ferroin indicator solution, iron 2+ , tris 1,10-phenanthroline-.kappa.n1,.kappa.n10-, oc-6-11-, sulfate 1:1, ferroin sulfate, ferroin solution, 1,10-phenanthroline ferrous sulfate, tris 1,10-phenanthroline-n1,n10 iron sulphate, ferrous 1,10-phenanthroline sulfate, ferroin indicator solution, 25 mmol/l, iron 2+ , tris 1,10-phenanthroline-kappan1,kappan10-, oc-6-11-, sulfate 1:1 |
PubChem CID | 84567 |
SMILES | [Fe++].[O-]S([O-])(=O)=O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15614240
|
Honeywell-Fluka
34531-100ML |
100 mL |
881.14 SEK
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15624240
|
Honeywell-Fluka
34531-500ML |
500 mL |
3170.38 SEK
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15634240
|
Honeywell-Fluka
34531-6X100ML |
6 x 100 mL | |||||||
15644240
|
Honeywell-Fluka
34531-6X500ML |
6 x 500 mL | |||||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
14634-91-4 | |
692.53 | |
CIWXFRVOSDNDJZ-UHFFFAOYSA-L | |
84567 |
C36H24FeN6O4S | |
MFCD00036428 | |
ferroin, ferroin indicator solution, iron 2+ , tris 1,10-phenanthroline-.kappa.n1,.kappa.n10-, oc-6-11-, sulfate 1:1, ferroin sulfate, ferroin solution, 1,10-phenanthroline ferrous sulfate, tris 1,10-phenanthroline-n1,n10 iron sulphate, ferrous 1,10-phenanthroline sulfate, ferroin indicator solution, 25 mmol/l, iron 2+ , tris 1,10-phenanthroline-kappan1,kappan10-, oc-6-11-, sulfate 1:1 | |
[Fe++].[O-]S([O-])(=O)=O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1 |
Specifications
14634-91-4 | |
[Fe(C12H8N2)3]SO4 | |
UN3082 | |
CIWXFRVOSDNDJZ-UHFFFAOYSA-L | |
iron(2+);1,10-phenanthroline;sulfate | |
84567 | |
Ferroin indicator solution |
C36H24FeN6O4S | |
MFCD00036428 | |
ferroin, ferroin indicator solution, iron 2+ , tris 1,10-phenanthroline-.kappa.n1,.kappa.n10-, oc-6-11-, sulfate 1:1, ferroin sulfate, ferroin solution, 1,10-phenanthroline ferrous sulfate, tris 1,10-phenanthroline-n1,n10 iron sulphate, ferrous 1,10-phenanthroline sulfate, ferroin indicator solution, 25 mmol/l, iron 2+ , tris 1,10-phenanthroline-kappan1,kappan10-, oc-6-11-, sulfate 1:1 | |
[Fe++].[O-]S([O-])(=O)=O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1.C1=CN=C2C(C=CC3=CC=CN=C23)=C1 | |
692.53 | |
692.52g/mol |
Safety and Handling
Product Identifier
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Ferroin indicator solution
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : 1,10-Phenanthrolinemonohydrochloride monohydrate