Learn More
FGF1 Polyclonal Antibody, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
2205.00 SEK - 19580.00 SEK
Specifications
Antigen | FGF1 |
---|---|
Concentration | 0.1-1.0 mg/mL |
Applications | ELISA, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17895383
|
Invitrogen
500-P17-50UG |
50 μg |
2205.00 SEK
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17823913
|
Invitrogen
500-P17-100UG |
100 μg |
2830.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17844423
|
Invitrogen
500-P17-1MG |
1 mg |
19580.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AA Sequence of recombinant protein: MFNLPPGNYK KPKLLYCSNG GHFLRILPDG TVDGTRDRSD QHIQLQLSAE SVGEVYIKST ETGQYLAMDT DGLLYGSQTP NEECLFLERL EENHYNTYIS KKHAEKNWFV GLKKNGSCKR GPRTHYGQKA ILFLPLPVSS D. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human FGF-acidic. Anti-Human FGF-acidic-specific antibody was purified by affinity chromatography employing an immobilized Human FGF-acidic matrix. Sandwich ELISA: To detect Human FGF-acidic by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human FGF-acidic (500-P17Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human FGF-acidic. Western Blot: To detect Human FGF-acidic by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Human FGF-acidic is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
FGF1 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein functions as a modifier of endothelial cell migration and proliferation, as well as an angiogenic factor. It acts as a mitogen for a variety of mesoderm- and neuroectoderm-derived cells in vitro, thus is thought to be involved in organogenesis.Specifications
FGF1 | |
ELISA, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P05230 | |
2246 | |
E.coli-derived Recombinant Human FGF-acidic | |
Antigen affinity chromatography | |
FGF1 |
0.1-1.0 mg/mL | |
Polyclonal | |
Lyophilized | |
RUO | |
PBS with no preservative | |
acidic eye-derived fibroblast growth factor; acidic eye-derived growth factor II; Acidic fibroblast growth factor; aFGF; beta-endothelial cell growth factor; Dffrx; ECGF; ECGFA; ECGFB; ECGF-beta; EDGF; EDGF II; Endothelial cell growth factor; Endothelial cell growth factor alpha; Endothelial cell growth factor beta; endothelial cell growth factor beta and alpha chains; endothelial cell growth factor, alpha; endothelial cell growth factor, beta; endothelial growth factor; Fam; FGF; FGF1; Fgf-1; FGF1 protein; Fgfa; FGF-alpha; Fibroblast growth factor; Fibroblast growth factor 1; fibroblast growth factor 1 (acidic); Fibroblast growth factor 1 (heparin binding); fibroblast growth factor, acidic; GLIO703; HBGF1; HBGF-1; heparin-binding growth factor; Heparin-binding growth factor 1; H-FGF-a; M-FGF-a; Prostatropin; unnamed protein product | |
FGF1 | |
Primary | |
-20°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.