Learn More
Eurofins DiscoverX FGFR3 Protein, Active
Active, recombinant human FGFR residues 447-761, containing an N-terminal His6-tag, for use in Kinase Assays.
5796.39 SEK - 44240.30 SEK
Specifications
Formulation | mg/mL in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
---|---|
Molecular Weight (g/mol) | 36.9 kDa |
Species | Human |
Recombinant | Recombinant |
Protein Tag | N-terminal 6His |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17380204
|
Eurofins DiscoverX
14-464 |
10 μg |
5796.39 SEK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17390204
|
Eurofins DiscoverX
14-464M |
250 μg |
44240.30 SEK
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fibroblast growth factor (FGF) superfamily consists of 23 members, all of which contain a conserved 120 amino acid (aa) core region that contains six identical, interspersed amino acids. Based on the pI factor, two main groups of FGF are named: acidic FGF (aFGF) and basic FGF (bFGF). The superfamily members act extracellularly through four tyrosine kinase FGF receptors (FGR1 through FGR4), with multiple specificities noted for almost all FGFs. Biological binding of aFGF to its receptors will result in phosphorylation of the receptor and non-receptor proteins and generate its function. FGR3 (FGFR-3, Fibroblast growth factor receptor 3, EC 2.7.1.112) is expressed mainly in bone, central nervous system and tumors. Inhibition of FGR3 may be useful in the treatment of dwarfism as well as some tumors.
GenBank M58051
Specifications
mg/mL in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. | |
Human | |
N-terminal 6His | |
Truncated | |
FGFR3, active |
36.9 kDa | |
Recombinant | |
Recombinant enzyme expressed in SF21 insect cells | |
≥60% |