Specifications
Molecular Weight (g/mol) | 372.09 |
Purity | ≥97.5 % |
Quantity | 100 mg |
Color | White to beige |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0435993 | Certificate of Analysis | 26/01/2022 |
Product Content Correction
Product Title
Fialuridine, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction