missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibrillin 2 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
1935.00 SEK - 4555.00 SEK
Specifications
Antigen | Fibrillin 2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18332334
|
Bio-Techne
NBP3-03631-20UL |
20 μL |
1935.00 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18391046
|
Bio-Techne
NBP3-03631-100UL |
100 μL |
4555.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fibrillin 2 Polyclonal specifically detects Fibrillin 2 in Human, Mouse samples. It is validated for Western Blot.Specifications
Fibrillin 2 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
CCA, congenital contractural arachnodactyly, DA9, fibrillin 2, fibrillin 5, fibrillin-2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 2733-2912 of human Fibrillin 2 (NP_001990.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Cell Biology, Cellular Markers | |
PBS with 50% glycerol, pH7.3. | |
2201 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title