missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibronectin Antibody (A17), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-23524AF488
This item is not returnable.
View return policy
Description
Fibronectin Monoclonal antibody specifically detects Fibronectin in Human, Bovine, Chicken samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)Specifications
Fibronectin | |
Monoclonal | |
Alexa Fluor 488 | |
CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF | |
Lysed bovine corneal endothelial cells and extracellular matrix | |
100 μg | |
Cancer, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
2335 | |
Store at 4°C in the dark. | |
IgG1 |
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
A17 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Bovine, Chicken | |
Purified |