Learn More
Hettich Lab Technology™ Filter Cards
Filter Cards are used for collecting cyto centrifuge samples and come in sizes to accommodate a variety of chambers and carriers.
Brand:  Hettich Lab Technology™ 1696
Additional Details : Weight : 0.09000kg
Product Code. 10530772
  • 373.00 SEK / Pack of 100
Estimated Shipment: 05-08-2024
to see stock.
Description
Description
  • For cyto centrifuge systems and cell evaluations
  • Suitable for a variety of cyto chambers and slide carriers
Specifications
Specifications
Filter Card