missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Filter Funnel

Filter funnel available in a buchner shape or a conical shape.

Brand:  ROBU VitraPOR™ 22251

Additional Details : Weight : 0.33720kg

 View more versions of this product

Product Code. 16813046

  • 2735.00 SEK / Pack of 10
Estimated Shipment: 06-09-2024
to see stock.
Add to basket

Description

Description

  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications

Specifications

25 mL
Conical Shape
10/Pk.
Borosilicate Glass (3.3)
1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers