Learn More
Kartell™ Dispolab™ Polyethylene Finned Plug
Designed for use with disposable tubes.
Brand: Kartell™ 8830504
224.07 SEK valid until 2024-08-31
Use promo code "21641" to get your promotional price.
Additional Details : Weight : 0.68400kg
Description
Designed for use with disposable tubes.
- Made from polyethylene
- Available in different colors
Specifications
Finned Plug | |
Polyethylene | |
15 to 17 mm Tubes |
CE-IVD | |
Blue |