Learn More
Fisherbrand™ 3/8 in. Dot Labels
GREENER_CHOICE
Fisher branded Cryogenic Storage Labels use a special adhesive that is designed to withstand extreme cold temperatures as low as -196°C (-311°F). Our cryo labels can be printed/written on and will not bleed, smudge or smear–perfect for identifying specimens after long periods of cryogenic storage.
Brand:  Fisherbrand™ 11010235
150.50 SEK valid until 2024-12-31
Use promo code "21676" to get your promotional price.
 View more versions of this product
Product Code. 17867221
  • 175.00 SEK / Each
Estimated Shipment: 13-03-2025
to see stock.
Explore more special offers
Description
  • Labels can be printed/written on and will not bleed, smudge or smear - perfect for identifying specimens after long periods of cryogenic storage
Specifications
Specifications
Red
Non-sterile
0.37 in.
Dot label roll
1000/Roll
9.55 mm