Learn More
Fisherbrand™ Accumet™ Temperature-Compensated Two-Cell Conductivity Probes
Fisherbrand™ Accumet™ Temperature-Compensated Two-Cell Conductivity Probes - Mercury - Free, k=1, 20μS to 20 mS/cm, with 8 pin connector
Brand: Fisherbrand™ 93X219050
3899.80 SEK valid until 2024-12-15
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
Fisherbrand™ Accumet™ Temperature-Compensated Two-Cell Conductivity Probes - Mercury - Free, k=1, 20μS to 20 mS/cm, with 8 pin connector
Description
For use on Accumet™ Excel (XL) series benchtop meters
- Immersion-type cell
- Use in standard electrode holder: unique compact size is same as typical pH electrode
- Built-in temperature sensors provide temperature compensated readings
- Epoxy body
Specifications
1 | |
Conductivity | |
Conductivity Probe | |
20 μS to 20 mS/cm |
8-pin | |
Temperature Compensated | |
Epoxy Body |