Learn More
Fisherbrand™ Accumet™ Temperature-Compensated Two-Cell Conductivity Probes
Fisherbrand™ Accumet™ Temperature-Compensated Two-Cell Conductivity Probes - Mercury - Free, k=0.1, 0.5 to 2,000μS/cm, with 8 pin connector
Brand: Fisherbrand™ 93X219051
4167.06 SEK valid until 2024-12-15
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
Fisherbrand™ Accumet™ Temperature-Compensated Two-Cell Conductivity Probes - Mercury - Free, k=0.1, 0.5 to 2,000μS/cm, with 8 pin connector
Description
For use on Accumet™ Excel (XL) series benchtop meters
- Immersion-type cell
- Use in standard electrode holder: unique compact size is same as typical pH electrode
- Built-in temperature sensors provide temperature compensated readings
- Epoxy body
Specifications
0.1 | |
Conductivity | |
Conductivity Probe | |
0.5 to 2000 μS/cm |
8-pin | |
Temperature Compensated | |
Epoxy Body |