Learn More
Fisherbrand™ Polypropylene Tubing Equal Angle Y-Shaped Connectors
Description
- Excellent chemical resistance
- Autoclavable polypropylene
- Serrated connecting surfaces to prevent tubing slipping
- Withstands flows to 20 p.s.i.
- The elasticity of the tubing will influence choice of size
- Dimensions listed are nominal
Specifications
Equal Angle Tubing Connector | |
0.125 to 0.219 in. | |
1.4 in. | |
4 to 6mm | |
0.2 in. | |
30.48 mm | |
White |
Autoclavable | |
Polypropylene | |
35.5 mm | |
5.08 mm | |
Y | |
1.2 in. | |
20 psi |