missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polyvinyl-Lined Polypropylene Caps

Product Code. 11583802
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Color White
Description Screw cap
Material HDPE
No. per Case 576/Cs.
Quantity 576/PK.
Sterility Sterile
Liner Material Polyvinyl
Neck Finish 43-400
Certifications/Compliance ISO 9001
For Use With (Equipment) Bottle
Closure Liner Polyvinyl
Closure Material Plastic
Closure Size 43-400 mm
Closure Type Screw Cap
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'