Learn More
Fisherbrand™ SureOne™ Reload Empty Racks
For use with Fisherbrand SureOne universal pipet tips
381.00 SEK - 388.00 SEK
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
11993456
![]() |
Fisherbrand™
T1000ER-FIS |
1250 μL |
388.00 SEK
Pack of 10 |
In Stock
Log in to see stock. |
||||||
11983456
![]() |
Fisherbrand™
T200ER-FIS |
200 to 300 μL |
382.00 SEK
Pack of 10 |
In Stock
Log in to see stock. ![]() |
||||||
11973456
![]() |
Fisherbrand™
T10ER-FIS |
10 to 20 μL |
381.00 SEK
Pack of 10 |
In Stock
Log in to see stock. |
||||||
Description
- Inserts (not included) sit flush with rack, allowing easy multichannel pipetting
- Notched clasp designed to prop lid open for autoclave venting
- Sure stack lid rim prevents racks from sliding around when stacked and carted around a lab
- Safe lock latch allows for one-handed opening and closing ability—won't pop open when dropped
- Sure snap design securely holds insert in place, preventing it from being lifted out while pipetting
- Flip-lid can be either a hinged or lift-off lid; hinged format does not prevent use of a multichannel pipet
Order Info
Inserts not included; must be ordered separately.
Specifications
Empty Rack | |
Pipette Tips |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ SureOne™ Reload Empty Racks