Learn More
Fisherbrand™ Visitorspec Safety Glasses, Coated, Antifog
Click to view available options
Coated lens
Description
Ideal for protecting visitors in industrial or medical settings.
- Economical safety spectacles
- 100% impact-resistant polycarbonate wraparound unilens design with blue temples
- Clear lens absorbs >99.9% of harmful UV light
- Fit over prescription eyeglasses
- Scratch-resistant, Anti-fog, Anti-static
Compliance
Meet ANSI™ Z87.1-2010 standards.
Specifications
Specifications
Certifications/Compliance | Meets ANSI Z87.1-2003 standards. |
Shape | Rectangle |
Lens Coating | Anti-fog, Anti-static, Scratch-resistant |
Lens Tint | Clear |
Product Type | Safety Glasses |
Frame Color | Clear |
Temple Style | Adjustable-length; spatula |
Material | Polycarbonate (Lens) |
Lens Material | Polycarbonate |
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Product Title
Fisherbrand™ Visitorspec Safety Glasses, Coated, Antifog >
Spot an opportunity for improvement?Share a Content Correction