Learn More
Cole-Parmer™ Fixed-Position Utility Clamps
Used to hold apparatus at a fixed 90 degree angle near the lab frame where no adjustment is required after set-up. Cole-Parmer™ Fixed-Position Utility Clamps are ideal for precise alignment of burettes, thermometers and reflux condensers.
Brand:  Cole-Parmer™ 08022-00
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11765399
  • 545.00 SEK / Each
Estimated Shipment: 27-09-2024
to see stock.
Add to basket
Description
Description
Clamps are supplied with non-slip vinyl sleeves or for temperatures above 100°C, fiberglass covers. Choose single adjustment style for standard use where one prong or “finger” is adjusted using a wing nut, while the other prong remains stationary or for a more versatile clamp choose the dual adjustment style. Dual adjustment clamps offer greater flexibility over a wider range. Utilizing thumbscrews, prongs are easily adjusted over a wider range. Clamps include an integral connector which accepts rods up to 0.75 in. (19mm) in diameter. An additional holder must be purchased in order to attach clamps to lab-frames, support stands or other apparatus.
Specifications
Specifications
1 Ea.
Nickel-plated Zinc
Used to hold apparatus at a fixed 90° angle near the lab frame where no adjustment is required after set-up
13.3
Utility Clamp
Burette
77 mm
Medium