missing translation for 'onlineSavingsMsg'
Learn More

Flash Point Standard TAG ASTM D56 (nominal value: 43.1 °C), Paragon Scientific™

Flash Point Certified Reference Material for the calibration and verification of laboratory instruments used in testing petroleum and derivative products for TAG Flash Point ASTM D56

Brand:  Paragon Scientific CRM-TAKR

Additional Details : CAS Number : 91770-15-9 Weight : 0.70000kg Transport : UN number : 1863 Chem class : 3 Pack group : III

Product Code. 16080040

  • 2800.00 SEK / 250mL
Estimated Shipment: 04-10-2024
to see stock.

Description

Description

  • Manufactured in accordance with ASTM / EN or IP test methods
  • Certified by Round Robin Method by independant test laboratories
  • Ensures good interlaboratory correlation
  • Supplied in tamper evident UN compliant security pack
  • Pack size 250 mL
  • 12 months shelf life
Specifications

Specifications

250 mL
UN 1223
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Nominal values quoted. Certified Values will vary from batch to batch.